missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Delangin Isoform A Monoclonal Antibody (KT54)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA172534
This item is not returnable.
View return policy
Description
Delangin Isoform A Monoclonal Antibody for ICC/IF, IP, Western Blot
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
Specifications
| Delangin Isoform A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q6KC79 | |
| NIPBL | |
| Delangin peptide coupled to BSA. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| KT54 | |
| Unconjugated | |
| NIPBL | |
| 4921518A06Rik; 4933421G18Rik; C79399; CDLS; CDLS1; Delangin; delangin homolog; ID; IDN3; IDN3-B; NIPBL; NIPBL, cohesin loading factor; Nipped-B homolog; Nipped-B homolog (Drosophila); nipped-B-like protein; Scc2; SCC2 homolog; sister chromatid cohesion 2 homolog | |
| Rat | |
| Protein G | |
| RUO | |
| 25836 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction