missing translation for 'onlineSavingsMsg'
Learn More

Delafloxacin meglumine, MedChemExpress

Product Code. p-200097399
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496556

Brand: Medchem Express HY14814A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Delafloxacin meglumine (ABT492 meglumine; RX-3341 meglumine; WQ-3034 meglumine) is a broad-spectrum fluoroquinolone antibiotic. Delafloxacin has a broad spectrum of activity that includes drug-resistant Staphylococcus aureus, Streptococcus pneumoniae, and Klebsiella pneumonia.

Delafloxacin meglumine (ABT492 meglumine

Specifications

Chemical Name or Material Delafloxacin meglumine
Physical Form Solid
CAS 352458-37-8
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H29ClF3N5O9
For Use With (Application) COVID-19-immunoregulation
Synonym ABT492 meglumine RX-3341 meglumine WQ-3034 meglumine
Solubility Information H2O : 50 mg/mL (78.62 mM; Need ultrasonic) ∣DMSO : ≥ 6.4 mg/mL (10.06 mM)
SMILES O=C(C1=CN(C2=NC(N)=C(F)C=C2F)C3=C(C=C(F)C(N4CC(O)C4)=C3Cl)C1=O)O.O[C@H]([C@H]([C@@H]([C@@H](CO)O)O)O)CNC
Molecular Weight (g/mol) 635.97
Formula Weight 635.97
Percent Purity 99.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.