missing translation for 'onlineSavingsMsg'
Learn More

Dehydrocostus Lactone, TRC

Product Code. 30350028
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350028 250 mg 250mg
30364445 100 mg 100mg
30360583 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350028 Supplier Toronto Research Chemicals Supplier No. TRCD229255250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-43-0
Molecular Formula C15 H18 O2
Molecular Weight (g/mol) 230.3
Synonym Azuleno[4,5-b]furan-2(3H)-one, decahydro-3,6,9-tris(methylene)-, (3aS,6aR,9aR,9bS)-, Azuleno[4,5-b]furan-2(3H)-one, decahydro-3,6,9-tris(methylene)-, [3aS-(3aα,6aα,9aα,9bβ)]-, Costus lactone, dehydro- (6CI,7CI), Guaia-4(15),10(14),11(13)-trien-12-oic acid, 6α-hydroxy-, γ-lactone (8CI), (-)-Dehydrocostus lactone, (3aS,6aR,9aR,9bS)-Decahydro-3,6,9-tris(methylene)azuleno[4,5-b]furan-2(3H)-one, Dehydrocostus lactone, Epiligulyl oxide
IUPAC Name (3aS,6aR,9aR,9bS)-3,6,9-trimethylidene-3a,4,5,6a,7,8,9a,9b-octahydroazuleno[4,5-b]furan-2-one
SMILES C=C1CC[C@@H]2[C@H](OC(=O)C2=C)[C@@H]3[C@H]1CCC3=C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H18O2/c1-8-4-7-12-10(3)15(16)17-14(12)13-9(2)5-6-11(8)13/h11-14H,1-7H2/t11-,12-,13-,14-/m0/s1
Formula Weight 230.1307
Chemical Name or Material Dehydrocostus lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.