missing translation for 'onlineSavingsMsg'
Learn More

Dehydrocostus Lactone, MedChemExpress

Product Code. 16459028
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459028 10 mg 10mg
16441448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16459028 Supplier Medchem Express Supplier No. HYN0591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dehydrocostus Lactone is a major sesquiterpene lactone isolated from the roots of Saussurea lappa.

Dehydrocostus Lactone is a major sesquiterpene lactone isolated from the roots of Saussurea lappa.

Specifications

Chemical Name or Material Dehydrocostus Lactone
CAS 477-43-0
Color White
Purity Grade Notes Research
Molecular Formula C15H18O2
Quantity 10 mg
Synonym (-)-Dehydrocostus lactone Epiligulyl oxide
Solubility Information DMSO : 250 mg/mL (1085.54 mM; Need ultrasonic) ∣Ethanol : 15 mg/mL (65.13 mM; Need ultrasonic)
SMILES O=C(O[C@@]([C@@]1([H])C(CC[C@@]21[H])=C)([H])[C@@]3([H])CCC2=C)C3=C
Molecular Weight (g/mol) 230.3
Formula Weight 230.3
Percent Purity 98.61%
Grade Research
Physical Form Crystals
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.