missing translation for 'onlineSavingsMsg'
Learn More

Dehydrocorydaline chloride, MedChemExpress

Product Code. 16499058
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499058 10 mg 10mg
16451478 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499058 Supplier Medchem Express Supplier No. HYN0674A
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid that regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. Dehydrocorydaline chloride elevates p38 MAPK activation. Anti-inflammatory and anti-cancer activities. Dehydrocorydaline chloride shows strong anti-malarial effects (IC50 =38 nM), and low cytotoxicity (cell viability > 90%) using P. falciparum 3D7 strain.

Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid that regulates protein expression of Bax, Bcl-2

Specifications

Chemical Name or Material Dehydrocorydaline chloride
Physical Form Solid
CAS 10605-03-5
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H24ClNO4
For Use With (Application) Cancer-Kinase/protease
Synonym 13-Methylpalmatine chloride
Solubility Information DMSO : 25 mg/mL (62.21 mM; Need ultrasonic)
SMILES CC1=C(C=CC(OC)=C2OC)C2=C[N+]3=C1C4=CC(OC)=C(OC)C=C4CC3.[Cl-]
Molecular Weight (g/mol) 401.88
Formula Weight 401.88
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.