missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydrocholic acid, MedChemExpress
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
Dehydrocholic acid is a synthetic bile acid, manufactured by the oxidation of cholic acid.
Specifications
Specifications
| Chemical Name or Material | Dehydrocholic acid |
| CAS | 81-23-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H34O5 |
| Quantity | 1 g |
| Solubility Information | DMSO : 25 mg/mL (62.11 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.66 mM; Need ultrasonic) |
| SMILES | C[C@H](CCC(O)=O)[C@H]([C@]12C)CC[C@@]1([H])[C@]3([H])C(C[C@]4([H])CC(CC[C@]4(C)[C@@]3([H])CC2=O)=O)=O |
| Molecular Weight (g/mol) | 402.52 |
| Formula Weight | 402.52 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction