missing translation for 'onlineSavingsMsg'
Learn More

Dehydrocholate sodium, MedChemExpress

Product Code. 16481088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481088 Supplier Medchem Express Supplier No. HYB0998

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dehydrocholic sodium is a hydrocholeretic, increasing bile output to clear increased bile acid load.

Dehydrocholic sodium is a hydrocholeretic, increasing bile output to clear increased bile acid load.

Specifications

Chemical Name or Material Dehydrocholate sodium
Physical Form Solid
CAS 145-41-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C24H33NaO5
Synonym Sodium dehydrocholate
Solubility Information H2O : 50 mg/mL (117.78 mM; Need ultrasonic) ∣DMSO : 20.83 mg/mL (49.07 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@H](CCC(O[Na])=O)[C@H]([C@]12C)CC[C@@]1([H])[C@]3([H])C(C[C@]4([H])CC(CC[C@]4(C)[C@@]3([H])CC2=O)=O)=O
Molecular Weight (g/mol) 424.51
Formula Weight 424.51
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.