missing translation for 'onlineSavingsMsg'
Learn More

Dehydroascorbic acid, MedChemExpress

Product Code. 16440697
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16440697 Supplier Medchem Express Supplier No. HY110281
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dehydroascorbic acid, a blood-brain barrier transportable form of vitamin C, mediates potent cerebroprotection in experimental stroke.

Dehydroascorbic acid, a blood-brain barrier transportable form of vitamin C, mediates potent cerebroprotection in experimental stroke.

Specifications

Chemical Name or Material Dehydroascorbic acid
CAS 490-83-5
Color Light Nude
Purity Grade Notes Research
Molecular Formula C6H6O6
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (287.17 mM; Need ultrasonic) ∣H2O : 5 mg/mL (28.72 mM; Need ultrasonic)
SMILES OC[C@@H]([C@](O1)([H])C(C(C1=O)=O)=O)O
Molecular Weight (g/mol) 174.11
Formula Weight 174.11
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.