missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydroaripiprazole, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Dehydroaripiprazole
Specifications
Specifications
| Chemical Name or Material | Dehydro Aripiprazole |
| CAS | 129722-25-4 |
| Analyte or Component Names | Dehydro Aripiprazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H25 Cl2 N3 O2 |
| Synonym | 7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1 H)-one, Dehydroaripiprazole, Aripiprazole Related Compound G, Aripiprazole Imp E (EP), Aripiprazole Imp. E (EP), Aripiprazole USP Related Compound G, Aripiprazole USP RC G, Aripiprazole Related Compound G, Aripiprazole Impurity E |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Clc1cccc(N2CCN(CCCCOc3ccc4C=CC(=O)Nc4c3)CC2)c1Cl |
| IUPAC Name | 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1 H-quinolin-2-one |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction