missing translation for 'onlineSavingsMsg'
Learn More

Dehydroandrographolide, MedChemExpress

Product Code. 16494867
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494867

Brand: Medchem Express HYN0676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dehydroandrographolide is extracted from herbal medicine Andrographis paniculata Nees. Dehydroandrographolide reduces oxidative stress in LPS-induced acute lung injury by inactivating iNOS. Dehydroandrographolide has anti-infective activity.

Dehydroandrographolide is extracted from herbal medicine Andrographis paniculata Nees. Dehydroandrographolide reduces oxidative stress in LPS-induced acute lung injury by inactivating iNOS. Dehydroandrographolide has anti-infective activity.

Specifications

Chemical Name or Material Dehydroandrographolide
CAS 134418-28-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H28O4
Quantity 50 mg
Solubility Information DMSO : 250 mg/mL (752.04 mM; Need ultrasonic)
SMILES C[C@@]([C@H]1C/C=C(C=CO2)/C2=O)(CC[C@H]3O)[C@@](CCC1=C)([H])[C@]3(C)CO
Molecular Weight (g/mol) 332.43
Formula Weight 332.43
Percent Purity 99.93%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.