missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic Acid, TRC

Product Code. 30367951
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367951 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367951 Supplier Toronto Research Chemicals Supplier No. TRCD2292351G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydroacetic Acid
CAS 520-45-6
Molecular Formula C8 H8 O4
Quantity 1 g
Synonym 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 3-Acetyl-6-methyl-1-oxacyclohex-5-ene-2,4-dione, 3-Acetyl-6-methyldihydropyrandione-2,4, 3-Acetyl-6-methylpyran-2,4(3H)-dione, 4-Hexenoic acid, 2-acetyl-5-hydroxy-3-oxo-, δ-lactone, Acetic acid, dehydro-, Biocide 470F, DHAA, DHS, Dehydracetic acid, Dehydroacetic acid, Geogard 111A, NSC 8770
InChI Formula InChI=1S/C8H8O4/c1-4-3-6(10)7(5(2)9)8(11)12-4/h3,7H,1-2H3
SMILES CC(=O)C1C(=O)OC(=CC1=O)C
IUPAC Name 3-acetyl-6-methylpyran-2,4-dione
Molecular Weight (g/mol) 168.15
Formula Weight 168.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.