missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 295.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 2.5 kg |
Solubility Information | Solubility in water: 330g/L water (25°C). Other solubilities: soluble in propylene glycol: 497.4g/l (25°C),methanol: 110.8g/l (25°C),insoluble in benzene,acetone |
Formula Weight | 190.13 |
Percent Purity | 99% |
Physical Form | Powder |
Show More |
Safety and Handling
Product Identifier
- Dehydroacetic acid sodium salt
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'