missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic acid, 98%

Product Code. 11463418
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463418 50 g 50g
11473418 250 g 250g
11453418 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463418 Supplier Thermo Scientific Alfa Aesar Supplier No. A17827.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 520-45-6 | C8H8O4 | 168.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-45-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00066709
InChI Key PGRHXDWITVMQBC-UHFFFAOYSA-N
Synonym dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
PubChem CID 122903
IUPAC Name 3-acetyl-6-methylpyran-2,4-dione
SMILES CC1=CC(=O)C(C(=O)O1)C(=O)C

Specifications

Melting Point 109°C to 113°C
Boiling Point 269°C to 270°C
Flash Point 157°C (314°F)
Quantity 50 g
Beilstein 6129
Merck Index 14,2865
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material Dehydroacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.