missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic acid 1000 μg/mL in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30321724
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321724 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321724 Supplier Dr. Ehrenstorfer Supplier No. DREA12114000EA1000
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydroacetic Acid
CAS 520-45-6
Analyte or Component Names Dehydroacetic Acid
Shipping Condition +20°C
Molecular Formula C8 H8 O4
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C8H8O4/c1-4-3-6(10)7(5(2)9)8(11)12-4/h3,7H,1-2H3
SMILES CC(=O)C1C(=O)OC(=CC1=O)C
IUPAC Name 3-acetyl-6-methylpyran-2,4-dione
Solvent or Matrix Ethyl Acetate
Molecular Weight (g/mol) 168.15
Quantity 1 mL
Formula Weight 168.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.