missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Nisoldipine, TRC

Product Code. 30354884
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354884 5 mg 5mg
30356913 50 mg 50mg
30358687 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354884 Supplier Toronto Research Chemicals Supplier No. TRCD2299005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103026-83-1
Molecular Formula C20 H22 N2 O6
Molecular Weight (g/mol) 386.4
Synonym 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(2-nitrophenyl)-, 3-methyl 5-(2-methylpropyl) ester, 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(2-nitrophenyl)-, methyl 2-methylpropyl ester (9CI), 3-Isobutyl 5-methyl 2,6-dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate, Dehydro Nisoldipine
IUPAC Name 5-O-methyl 3-O-(2-methylpropyl) 2,6-dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate
SMILES COC(=O)c1c(C)nc(C)c(C(=O)OCC(C)C)c1c2ccccc2[N+](=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H22N2O6/c1-11(2)10-28-20(24)17-13(4)21-12(3)16(19(23)27-5)18(17)14-8-6-7-9-15(14)22(25)26/h6-9,11H,10H2,1-5H3
Formula Weight 386.1478
Chemical Name or Material Dehydro Nisoldipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.