missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Lovastatin, TRC

Product Code. 30354131
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354131 25 mg 25mg
30350997 2.5 mg 2.50mg
30366481 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354131 Supplier Toronto Research Chemicals Supplier No. TRCD22968525MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109273-98-5
Molecular Formula C24 H34 O4
Molecular Weight (g/mol) 386.52
Synonym (1S,3R,7S,8S,8aR)-3,7-Dimethyl-8-[2-[(2R)-6-oxo-3,6-dihydro-2H-pyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydro-naphthalen-1-yl, Lovastatin Imp. C (EP), Dehydrolovastatin
IUPAC Name [(1S,3R,7S,8S,8aR)-3,7-dimethyl-8-[2-[(2R)-6-oxo-2,3-dihydropyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3CC=CC(=O)O3)[C@@H]12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H34O4/c1-5-16(3)24(26)28-21-14-15(2)13-18-10-9-17(4)20(23(18)21)12-11-19-7-6-8-22(25)27-19/h6,8-10,13,15-17,19-21,23H,5,7,11-12,14H2,1-4H3/t15-,16-,17-,19-,20-,21-,23-/m0/s1
Formula Weight 386.2457
Chemical Name or Material (1S,3R,7S,8S,8aR)-3,7-Dimethyl-8-[2-[(2R)-6-oxo-3,6-dihydro-2H-pyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydro-naphthalen-1-yl (2S)-2-Methylbutanoate (Dehydrolovastatin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.