missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydro Glycopyrrolate Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | Dehydro Glycopyrrolate Bromide |
| Analyte or Component Names | Dehydro Glycopyrrolate Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19H26NO3 • Br |
| Synonym | 3-(2-(cyclopent-1-en-1-yl)-2-hydroxy-2-phenylacetoxy)-1,1-dimethylpyrrolidin-1-ium bromide |
| InChI Formula | InChI=1S/C19H26NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-10,17,22H,6-7,11-14H2,1-2H3;1H/q+1;/p-1 |
| Solution Type | Neat |
| SMILES | C[N+]1(CCC(OC(C(C2=CCCC2)(O)C3=CC=CC=C3)=O)C1)C.[Br-] |
| IUPAC Name | 3-(2-(cyclopent-1-en-1-yl)-2-hydroxy-2-phenylacetoxy)-1,1-dimethylpyrrolidin-1-ium bromide |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?