missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Glycopyrrolate Bromide, TRC

Product Code. 30369329
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369329 50 mg 50mg
30381737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369329 Supplier Toronto Research Chemicals Supplier No. TRCD83599050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydro Glycopyrrolate Bromide
Analyte or Component Names Dehydro Glycopyrrolate Bromide
Shipping Condition Room Temperature
Molecular Formula C19H26NO3 • Br
Synonym 3-(2-(cyclopent-1-en-1-yl)-2-hydroxy-2-phenylacetoxy)-1,1-dimethylpyrrolidin-1-ium bromide
InChI Formula InChI=1S/C19H26NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-10,17,22H,6-7,11-14H2,1-2H3;1H/q+1;/p-1
Solution Type Neat
SMILES C[N+]1(CCC(OC(C(C2=CCCC2)(O)C3=CC=CC=C3)=O)C1)C.[Br-]
IUPAC Name 3-(2-(cyclopent-1-en-1-yl)-2-hydroxy-2-phenylacetoxy)-1,1-dimethylpyrrolidin-1-ium bromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 396.32
Quantity 50 mg
Formula Weight 395.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.