missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEGS1 Recombinant Rabbit Monoclonal Antibody (K01_1T94)

Rabbit Recombinant Monoclonal Antibody
£390.00 - £414.00
Specifications
| Antigen | DEGS1 |
|---|---|
| Clone | K01_1T94 |
| Concentration | 1.30 mg/mL |
| Content And Storage | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30828433
![]() |
Invitrogen™
MA563443 |
100 μL |
£390.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30830444
|
Invitrogen™
MA564902 |
50 μL |
£414.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DEGS1 Recombinant Monoclonal Antibody for Western Blot
The product of the DEGS1 gene is a member of the membrane fatty acid (sphingolipid delta 4 ) desaturase family. Reported to function by inserting double bonds at delta 4 position in fatty acids. DEGS1 is an intracellular enzyme. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulation biosynthetic processing of the EGF receptor. Two splice variants have been identified.Specifications
| DEGS1 | |
| 1.30 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 | |
| O15121 | |
| 8560 | |
| A synthetic peptide of human MLD. | |
| Antibody |
| K01_1T94 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| DEGS1 | |
| AA536663; Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1 (Drosophila); degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; degenerative spermatocyte-like protein RDES; Degs; DEGS1; DEGS-1; delta 4-desaturase, sphingolipid 1; Delta(4)-desaturase; delta(4)-desaturase, sphingolipid 1; DES1; Des-1; dihydroceramide desaturase; dihydroceramide desaturase 1; dihydroceramide desaturase-1; DSH1; FADS7; fb81d06; hypothetical protein LOC507290; im:6909319; Mdes; membrane fatty acid (lipid) desaturase; membrane lipid desaturase; MGC5079; MIG15; migration-inducing gene 15 protein; MLD; sphingolipid 1; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; sphingolipid delta(4)-desaturase DES1; wu:fa25h01; wu:fb81d06 | |
| DEGS1 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title