missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEGS1 Recombinant Rabbit Monoclonal Antibody (9D6X8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555488
This item is not returnable.
View return policy
Description
Immunogen Sequence: MGSRVSREDF EWVYTDQPHA DRRREILAKY PEIKSLMKPD PNLIWIIIMM VLTQLGAFYI VKDLDWKWVI FGAYAFGSCI NHSMTLAIHE IAHNAAFGNC.
The product of the DEGS1 gene is a member of the membrane fatty acid (sphingolipid delta 4 ) desaturase family. Reported to function by inserting double bonds at delta 4 position in fatty acids. DEGS1 is an intracellular enzyme. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulation biosynthetic processing of the EGF receptor. Two splice variants have been identified.
Specifications
| DEGS1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15121 | |
| DEGS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human MLD. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9D6X8 | |
| Unconjugated | |
| DEGS1 | |
| AA536663; Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1 (Drosophila); degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; degenerative spermatocyte-like protein RDES; Degs; DEGS1; DEGS-1; delta 4-desaturase, sphingolipid 1; Delta(4)-desaturase; delta(4)-desaturase, sphingolipid 1; DES1; Des-1; dihydroceramide desaturase; dihydroceramide desaturase 1; dihydroceramide desaturase-1; DSH1; FADS7; fb81d06; hypothetical protein LOC507290; im:6909319; Mdes; membrane fatty acid (lipid) desaturase; membrane lipid desaturase; MGC5079; MIG15; migration-inducing gene 15 protein; MLD; sphingolipid 1; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; sphingolipid delta(4)-desaturase DES1; wu:fa25h01; wu:fb81d06 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8560 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction