missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DEGS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30903-100ug
This item is not returnable.
View return policy
Description
DEGS1 Polyclonal antibody specifically detects DEGS1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| DEGS1 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| Cell migration-inducing gene 15 protein, Degenerative spermatocyte homolog 1, degenerative spermatocyte homolog 1, lipid desaturase (Drosophila), degenerative spermatocyte homolog, lipid desaturase (Drosophila), DEGS, Des-1, DES1degenerative spermatocyte homolog, lipid desaturase, EC 1.14, FADS7, membrane fatty acid (lipid) desaturase, Membrane lipid desaturase, MGC5079, migration-inducing gene 15 protein, MLDdihydroceramide desaturase, sphingolipid delta 4 desaturase, sphingolipid delta(4)-desaturase DES1 | |
| Synthetic peptide derived from the human DEGS1 protein | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| 8560 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction