missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEGS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £251.00
Specifications
| Antigen | DEGS1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633158
|
Novus Biologicals
NBP3-12689-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659397
|
Novus Biologicals
NBP3-12689-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DEGS1 Polyclonal antibody specifically detects DEGS1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| DEGS1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| PBS, pH7.0 | |
| 8560 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell migration-inducing gene 15 protein, Degenerative spermatocyte homolog 1, degenerative spermatocyte homolog 1, lipid desaturase (Drosophila), degenerative spermatocyte homolog, lipid desaturase (Drosophila), DEGS, Des-1, DES1degenerative spermatocyte homolog, lipid desaturase, EC 1.14, FADS7, membrane fatty acid (lipid) desaturase, Membrane lipid desaturase, MGC5079, migration-inducing gene 15 protein, MLDdihydroceramide desaturase, sphingolipid delta 4 desaturase, sphingolipid delta(4)-desaturase DES1 | |
| Produced in rabbits immunized with E. coli-derived Human DEGS1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title