missing translation for 'onlineSavingsMsg'
Learn More

Defluoro N-Benzyl Paroxetine Hydrochloride, TRC

Product Code. 30366625
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366625 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366625 Supplier Toronto Research Chemicals Supplier No. TRCD22905550MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Defluoro N-Benzyl Paroxetine Hydrochloride
CAS 105813-39-6
Purity Grade Notes HPLC
Molecular Formula C26H28ClNO3
Quantity 50 mg
Synonym trans-(-)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenyl-1-(phenylmethyl)piperidine Hydrochloride
InChI Formula InChI=1S/C26H27NO3.ClH/c1-3-7-20(8-4-1)16-27-14-13-24(21-9-5-2-6-10-21)22(17-27)18-28-23-11-12-25-26(15-23)30-19-29-25;/h1-12,15,22,24H,13-14,16-19H2;1H/t22-,24-;/m0./s1
SMILES C1CN(C[C@H]([C@@H]1C2=CC=CC=C2)COC3=CC4=C(C=C3)OCO4)CC5=CC=CC=C5.Cl
Molecular Weight (g/mol) 437.96
Formula Weight 437.1758
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.