missing translation for 'onlineSavingsMsg'
Learn More

Deflazacort, MedChemExpress

Product Code. 16471927
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471927 50 mg 50mg
16466246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471927 Supplier Medchem Express Supplier No. HY13609
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Deflazacort, a glucocorticoid, is an inactive prodrug and is converted rapidly to the active metabolite 21-desacetyldeflazacort. Deflazacort is used as an anti-inflammatory and immunosuppressant.

Deflazacort, a glucocorticoid, is an inactive prodrug and is converted rapidly to the active metabolite 21-desacetyldeflazacort. Deflazacort is used as an anti-inflammatory and immunosuppressant.

Specifications

Chemical Name or Material Deflazacort
CAS 14484-47-0
Color White
Purity Grade Notes Research
Molecular Formula C25H31NO6
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (226.49 mM)
SMILES O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])C[C@]5([H])[C@@]4(C(COC(C)=O)=O)N=C(C)O5)=C1
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.