missing translation for 'onlineSavingsMsg'
Learn More

Deferoxamine mesylate, MedChemExpress

Product Code. 16432236
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16432236

Brand: Medchem Express HYB0988

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Deferoxamine mesylate (Deferoxamine B mesylate) is an iron chelator (binds to Fe(III) and many other metal cations), is widely used to reduce iron accumulation and deposition in tissues.

Deferoxamine mesylate upregulates HIF-1α levels with good antioxidant activity. Deferoxamine mesylate also shows anti-proliferative activity, can induce apoptosis and autophagy in cancer cells. Deferoxamine mesylate can be used in studies of diabetes, neurodegenerative diseases as well as anti-cancer and anti-COVID-19.

Specifications

Chemical Name or Material Deferoxamine mesylate
CAS 138-14-7
Color White
Purity Grade Notes Research
Molecular Formula C26H52N6O11S
Quantity 500 mg
Synonym Desferrioxamine B mesylate DFOM
Solubility Information H2O : 250 mg/mL (380.64 mM; Need ultrasonic)
SMILES O=C(N(CCCCCN)O)CCC(NCCCCCN(C(CCC(NCCCCCN(C(C)=O)O)=O)=O)O)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 656.79
Formula Weight 656.79
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.