missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Deferoxamine mesylate

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Deferoxamine mesylate is an iron chelator. Also hypoxia mimetic and inhibitor of ferroptosis. Has neuroprotective effects in animal models of stroke, traumatic brain injury and neurodegenerative diseases.
Specifications
Specifications
| Chemical Name or Material | N4-[5-[[4-[[5-(Acetylhydroxyamino)pentyl]amino-1,4-dioxobutyl]hydroxyamino]pentyl]-N1-(5-aminopentyl)-N1-hydroxybutanediamide methanesulfonate |
| CAS | 138-14-7 |
| Quantity | 50 mg |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C25H48N6O8.CH4O3S |
| Formula Weight | 656.79 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction