missing translation for 'onlineSavingsMsg'
Learn More

Deferiprone, TRC

Product Code. 30382824
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382824 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382824 Supplier Toronto Research Chemicals Supplier No. TRCD47400025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deferiprone
CAS 30652-11-0
Molecular Formula C7 H9 N O2
Quantity 25 g
Synonym 3-Hydroxy-1,2-dimethyl-4(1H)-pyridinone, 4(1H)-Pyridone, 3-hydroxy-1,2-dimethyl- (8CI), 1,2-Dimethyl-3-hydroxy-4(1H)-pyridinone, 1,2-Dimethyl-3-hydroxy-4-pyridone, 1,2-Dimethyl-3-hydroxypyridin-4-one, 1,2-Dimethyl-3-hydroxypyridine-4-one, 3-Hydroxy-1,2-dimethyl-4(1H)-pyridinone, 3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, 3-Hydroxy-1,2-dimethyl-4-pyridinone, 3-Hydroxy-1,2-dimethyl-4-pyridone, APO 66, CGP 37391, CP 20, CP 20 (chelating agent), DN 18001AF, Deferione, Deferiprone, Ferinil, Ferriprox, L 1, L 1 (chelating agent), PL 1
InChI Formula InChI=1S/C7H9NO2/c1-5-7(10)6(9)3-4-8(5)2/h3-4,10H,1-2H3
SMILES CN1C=CC(=O)C(=C1C)O
IUPAC Name 3-hydroxy-1,2-dimethylpyridin-4-one
Molecular Weight (g/mol) 139.15
Formula Weight 139.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.