missing translation for 'onlineSavingsMsg'
Learn More

Deferasirox, TRC

Product Code. 30135552
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135552 100 mg 100mg
30012569 10 mg 10mg
30008003 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135552 Supplier Toronto Research Chemicals Supplier No. TRCD228650100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201530-41-8
Molecular Formula C21 H15 N3 O4
Molecular Weight (g/mol) 373.36
Synonym Benzoic acid, 4-[3,5-bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]-, 4-[3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoic acid, 4-[3,5-Bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoic acid, Asunra, Deferasirox, Exjade, ICL 670, ICL 670A, Jadenu
IUPAC Name 4-[3,5-bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoic acid
SMILES OC(=O)c1ccc(cc1)n2nc(nc2c3ccccc3O)c4ccccc4O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H15N3O4/c25-17-7-3-1-5-15(17)19-22-20(16-6-2-4-8-18(16)26)24(23-19)14-11-9-13(10-12-14)21(27)28/h1-12,25-26H,(H,27,28)
Formula Weight 373.1063
Chemical Name or Material Deferasirox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.