missing translation for 'onlineSavingsMsg'
Learn More

Deferasirox, Mikromol™

Product Code. 17982756
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982756 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17982756 Supplier Mikromol Supplier No. MM3527.00
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Deferasirox
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deferasirox
CAS 201530-41-8
Analyte or Component Names Deferasirox
Shipping Condition Room Temperature
Molecular Formula C21 H15 N3 O4
Certifications/Compliance ISO 17034
Synonym 4-[3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]-benzoic acid, 4-[3,5-Bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoic acid, 4-[3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]benzoic acid, Asunra, Deferasirox, Exjade, ICL 670, ICL 670A
InChI Formula InChI=1S/C21H15N3O4/c25-17-7-3-1-5-15(17)19-22-20(16-6-2-4-8-18(16)26)24(23-19)14-11-9-13(10-12-14)21(27)28/h1-12,25-26H,(H,27,28)
SMILES OC(=O)c1ccc(cc1)n2nc(nc2c3ccccc3O)c4ccccc4O
IUPAC Name 4-[3,5-bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoic acid
Molecular Weight (g/mol) 373.36
Quantity 250 mg
Formula Weight 373.1063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.