missing translation for 'onlineSavingsMsg'
Learn More

Defensin beta 1 Antibody (1F5), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen Defensin beta 1
Clone 1F5
Dilution Flow Cytometry 1:50-1:200, ELISA
Applications Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043552
View Documents
Novus Biologicals
NBP3-26625-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043733
View Documents
Novus Biologicals
NBP3-26625-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Defensin beta 1 Monoclonal antibody specifically detects Defensin beta 1 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications

Specifications

Defensin beta 1
Flow Cytometry 1:50-1:200, ELISA
Monoclonal
Purified
RUO
Human
BD1, BD-1, beta-defensin 1, beta-defensin-1, DEFB-1, DEFB101, defensin, beta 1HBD-1, hBD-1, HBD1MGC51822
A synthesized peptide derived from Human Defensin beta 1 [UniProt P60022]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
1F5
Flow Cytometry, ELISA
Unconjugated
Rabbit
Biologically Active Proteins
PBS, pH 7.4, 150mM NaCl and 50% glycerol
1672
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.