missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Defensin alpha 5 Antibody (8C8), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60002PE
This item is not returnable.
View return policy
Description
Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for Flow Cytometry.
Specifications
| Defensin alpha 5 | |
| Monoclonal | |
| PE | |
| Q01523 | |
| DEFA5 | |
| Full-length human Defensin alpha 5 [Swiss-Prot# Q01523]. The propeptide is the full-length protein. In the mature form, it is cleaved at residue 63 (alanine). | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for HD5. It does not cross-react with HD6, lysozyme or sPLA2. | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 8C8 | |
| Flow Cytometry | |
| DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728 | |
| Mouse | |
| 10 kDa | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 1670 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction