missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Defensin alpha-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592407
This item is not returnable.
View return policy
Description
Immunogen sequence: EPLQARADEV AAAPEQIAAD IPEVVVSLAW DESLAPKHPG SRKNMDCYCR IPACIAGERR YGTCIYQGRL WAFCC; Positive Samples: MCF-7, 22Rv1, SW480.
Defensin alpha-1 is part of a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. Defensins are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The Defensin alpha-1 protein is encoded by the DFA1 gene and is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8, and this gene differs from defensin, alpha 3 by only one amino acid. The DFA1 gene and the gene encoding defensin, alpha 3 are both subject to copy number variation. Two transcript variants encoding different isoforms of Defensin alpha-1 have been found.
Specifications
| Defensin alpha-3 | |
| Polyclonal | |
| Unconjugated | |
| DEFA3 | |
| DEF3; DEFA3; defensin 3, neutrophil-specific; defensin alpha 3; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; HNP-2; HNP3; HNP-3; HP 3-56; HP-2; HP3; HP-3; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1668 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59666 | |
| DEFA3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-94 of human DEFA3 (NP_0052081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction