missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Defensin 1/3 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA519228

Product Code. 11377203

  • £442.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 3 by only one amino acid. This gene and the gene encoding defensin, alpha 3 are both subject to copy number variation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Defensin 1/3
Polyclonal
Unconjugated
DEFA1
alpha-defensin 1; Alpha-defensin 3; cryptdin-1; DEF1; DEF3; Defa1; DEFA1B; DEFA2; Defa27; Defa3; Defcr; Defcr1; Defcr3; Defcr-3; defensin 3, neutrophil-specific; defensin alpha 1; defensin alpha 3; defensin related cryptdin peptide 1; defensin, alpha 1; defensin, alpha 1, myeloid-related sequence; defensin, alpha 2; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; defensin, alpha, 27; defensin, alpha, 3; defensin-1; defensin-related cryptdin peptide; Defensin-related cryptdin-3; HNP-1; HNP-2; HNP3; HNP-3; HP 1-56; HP 3-56; HP1; HP-1; HP-2; HP3; HP-3; MRS; myeloid-related sequence; neutrophil defensin 1; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3
Goat
Ammonium sulfate precipitation
RUO
1667, 1668
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P59665, P59666
DEFA1, DEFA3
Synthetic peptide sequence (DESLAPKHPGSRKN) corresponding to the internal amino acids of DEFA1/3.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.