missing translation for 'onlineSavingsMsg'
Learn More

DEFB126 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18352024 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352024 100 μL 100µL
18366876 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18352024 Supplier Novus Biologicals Supplier No. NBP303581100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DEFB126 Polyclonal specifically detects DEFB126 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB126
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias bA530N10.1, beta defensin 26, beta-defensin 126, Beta-defensin 26, C20orf8, DEFB26, DEFB-26chromosome 20 open reading frame 8, defensin, beta 126ESP13.2, Epididymal secretory protein 13.2, epididymal secretory protein ESP13.2, hBD-26
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-128 of rat Defb26 DEFB126 (NP_001032595.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81623
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.