missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEFB107A Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142805
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Has antibacterial activity.
Specifications
| DEFB107A | |
| Polyclonal | |
| Unconjugated | |
| DEFB107A | |
| BD-7; Beta-defensin 107; beta-defensin 7; DEFB107; DEFB107A; DEFB107B; DEFB7; DEFB-7; defensin beta 107A; Defensin, beta 107; defensin, beta 107A; defensin, beta 7 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 245910 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8IZN7 | |
| DEFB107A | |
| Peptide with sequence HRALISKRMEGHCE (aa 29-42). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction