missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Defactinib

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Defactinib is a potent and selective focal adhesion kinase (FAK) and proline rich tyrosine kinase 2 (Pyk2) inhibitor (IC50 = 0.6 nM at both). The compound shows >100-fold selectivity for FAK and Pyk2 over other kinases. Defactinib inhibits FAK phosphorylation in vivo (EC50 = 26 nM), and inhibits tumor growth. Defactinib also decreases the viability of thyroid cancer cells, showing greater selectivity for TT and K1 cells compared to FAK Inhibitor 14 (Cat. No. 3414) (IC50 values are 1.98 μM and 10.34 μM, respectively). Defactinib suppresses colony growth of Pnf cells when combined with Selumetinib (Cat. No. 6815).
Specifications
Specifications
| Chemical Name or Material | N-Methyl-4-[[4-[[[3-[methyl(methylsulfonyl)amino]-2-pyrazinyl]methyl]amino]-5-(trifluoromethyl)-2-pyrimidinyl]amino]benzamide |
| CAS | 1073154-85-4 |
| Quantity | 5 mg |
| Target | Focal Adhesion Kinase Inhibitors |
| Molecular Formula | C20H21F3N8O3S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction