missing translation for 'onlineSavingsMsg'
Learn More

Defactinib hydrochloride, MedChemExpress

Product Code. 16445806
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16445806

Brand: Medchem Express HY12289A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Defactinib hydrochloride (VS-6063 hydrochloride; PF 04554878 hydrochloride) is a novel FAK inhibitor, which inhibits FAK phosphorylation at the Tyr397 site in a time- and dose-dependent manner.

Defactinib hydrochloride (VS-6063 hydrochloride

Specifications

Chemical Name or Material Defactinib hydrochloride
CAS 1073160-26-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22ClF3N8O3S
Quantity 100 mg
Synonym VS-6063 hydrochloride PF 04554878 hydrochloride
Solubility Information DMSO : 30 mg/mL (54.85 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC)C1=CC=C(NC2=NC=C(C(F)(F)F)C(NCC3=NC=CN=C3N(C)S(=O)(C)=O)=N2)C=C1.[H]Cl
Molecular Weight (g/mol) 546.95
Formula Weight 546.95
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.