missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEFA6 Antibody (2D2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00001671-M01
This item is not returnable.
View return policy
Description
DEFA6 Monoclonal antibody specifically detects DEFA6 in Human samples. It is validated for Western Blot, ELISA
Specifications
| DEFA6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DEF6HD-6, defensin 6, Defensin, alpha 6, defensin, alpha 6, Paneth cell-specific, defensin-6 | |
| DEFA6 (NP_001917.1, 1 a.a. ~ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRTLTILTAVLLVALQAKAEPLQAEDDPLQAKAYEADAQEQRGANDQDFAVSFAEDASSSLRALGSTRAFTCHCRRSCYSTEYSYGTCTVMGINHRFCCL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2D2 | |
| Western Blot, ELISA | |
| NP_001917.1 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 1671 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction