missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Defensin 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582540
This item is not returnable.
View return policy
Description
Immunogen sequence: ESLQERADEA TTQKQSGEDN QDLAISFAGN GLSALRTSGS QARATCYCRT GRCATRESLS GVCEISGRLY RLCCR Highest antigen sequence indentity to the following orthologs: Rat - 49%, Mouse - 39%.
Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several of the alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 5, is highly expressed in the secretory granules of Paneth cells of the ileum.
Specifications
| Defensin 5 | |
| Polyclonal | |
| Unconjugated | |
| DEFA5 | |
| DEF5; DEFA5; defensin 5; defensin alpha 5; Defensin, alpha 5; defensin, alpha 5, Paneth cell-specific; defensin, alpha 5, preproprotein; defensin-5; HD5; HD-5; HD5(20-94); HD5(23-94); HD5(29-94); HD5(56-94); HD5(63-94); MGC129728 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1670 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01523 | |
| DEFA5 | |
| Recombinant protein fragment of Human DEFA5 (Product #RP-89636) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction