missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEFA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562362
This item is not returnable.
View return policy
Description
Immunogen sequence: SSALQVSGST RGMVCSCRLV FCRRTELRVG NCLIGGVSFT YCCTRVD Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 36%.
Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes are clustered on chromosome 8. This gene differs from other genes of this family by an extra 83-base segment that is apparently the result of a recent duplication within the coding region. The protein encoded by this gene, defensin, alpha 4, is found in the neutrophils; it exhibits corticostatic activity and inhibits corticotropin stimulated corticosterone production.
Specifications
| DEFA4 | |
| Polyclonal | |
| Unconjugated | |
| DEFA4 | |
| Alpha-defensin 4; corticostatin; Crp4; DEF4; Defa28; Defa4; Defcr4; defensin alpha 4; Defensin, alpha 4; defensin, alpha 4, corticostatin; defensin, alpha 4, preproprotein; defensin, alpha, 4; Defensin-related cryptdin-4; HNP-4; HP4; HP-4; Neutrophil defensin 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12838 | |
| DEFA4 | |
| Recombinant protein corresponding to Human Defensin. Recombinant protein control fragment (Product #RP-101423). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction