missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113458
This item is not returnable.
View return policy
Description
DEF6 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
Specifications
| DEF6 | |
| Polyclonal | |
| Unconjugated | |
| DEF6 | |
| 2410003F05Rik; 6430538D02Rik; AV094905; DEF6; DEF-6; DEF6 guanine nucleotide exchange factor; DEF6, guanine nucleotide exchange factor; differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; differentially expressed in FDCP 6 homolog (mouse); Ibp; IRF-4-binding protein; IRF4-binding protein; LOW QUALITY PROTEIN: differentially expressed in FDCP 6 homolog; OTTHUMP00000016251; RP3-329A5.5; SLAT; Slat2; Slat6; SWAP70L; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells | |
| Rabbit | |
| Protein G | |
| RUO | |
| 23853, 50619 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 8 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8C2K1, Q9H4E7 | |
| DEF6 | |
| Recombinant Human Differentially expressed in FDCP 6 homolog protein (20-114AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction