missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545672
This item is not returnable.
View return policy
Description
Peptide sequence: QERYIERAQQ EKEELQQEMA QQSRSLQQAQ QQLEEVRQNR QRADEDVEAA Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 83%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 90%; Rabbit: 93%; Rat: 93%.
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
Specifications
| DEF6 | |
| Polyclonal | |
| Unconjugated | |
| DEF6 | |
| 2410003F05Rik; 6430538D02Rik; AV094905; DEF6; DEF-6; DEF6 guanine nucleotide exchange factor; DEF6, guanine nucleotide exchange factor; differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; differentially expressed in FDCP 6 homolog (mouse); Ibp; IRF-4-binding protein; IRF4-binding protein; LOW QUALITY PROTEIN: differentially expressed in FDCP 6 homolog; OTTHUMP00000016251; RP3-329A5.5; SLAT; Slat2; Slat6; SWAP70L; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50619 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4E7 | |
| DEF6 | |
| Synthetic peptide directed towards the C-terminal of human DEF6 (aa 480-529). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction