Learn More
Invitrogen™ DEF6 Monoclonal Antibody (OTI1E3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526107
Product Code. 15945704
  • £474.00 / 100µL
Description
DEF6 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DEF6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C2K1, Q9H4E7
DEF6
Full length human recombinant protein of DEF6 produced in HEK293T cell.
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI1E3
Unconjugated
DEF6
2410003F05Rik; 6430538D02Rik; AV094905; DEF6; DEF-6; DEF6 guanine nucleotide exchange factor; DEF6, guanine nucleotide exchange factor; differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; differentially expressed in FDCP 6 homolog (mouse); Ibp; IRF-4-binding protein; IRF4-binding protein; LOW QUALITY PROTEIN: differentially expressed in FDCP 6 homolog; OTTHUMP00000016251; RP3-329A5.5; SLAT; Slat2; Slat6; SWAP70L; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells
Mouse
Affinity Chromatography
RUO
23853, 309642, 50619, 608163
-20°C, Avoid Freeze/Thaw Cycles