missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEF6 Antibody (OTI3F9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71868B
This item is not returnable.
View return policy
Description
DEF6 Monoclonal specifically detects DEF6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DEF6 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| DEF6 | |
| Full length recombinant protein of human DEF6 (NP_071330) produced in HEK293T cells. | |
| 0.1 mL | |
| 50619.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3F9 | |
| Western Blot, Immunohistochemistry | |
| DEF-6, differentially expressed in FDCP 6 homolog, differentially expressed in FDCP 6 homolog (mouse), IBPdifferentially expressed in FDCP (mouse homolog) 6, IRF4-binding protein, SLAT, SWAP70L, SWAP-70-like adaptor protein of T cells | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction