missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEF6 Antibody (1F2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00050619-M01
This item is not returnable.
View return policy
Description
DEF6 Monoclonal antibody specifically detects DEF6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
Specifications
| DEF6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 50619 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 1F2 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| DEF-6, differentially expressed in FDCP 6 homolog, differentially expressed in FDCP 6 homolog (mouse), IBPdifferentially expressed in FDCP (mouse homolog) 6, IRF4-binding protein, SLAT, SWAP70L, SWAP-70-like adaptor protein of T cells | |
| DEF6 (AAH17504, 1 a.a. ~ 336 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MALRKELLKSIWYAFTALDVEKSGKVSKSQLKVLSHNLYTVLHIPHDPVALEEHFRDDDDGPVSSQGYMPYLNKYILDKVEEGAFVKEHFDELCWTLTAKKNYRADSNGNSMLSNQDAFRLWCLFNFLSEDKYPLIMVPDEVEYLLKKVLSSMSLEVSLGELEELLAQEAQVAQTTGGLSVWQFLELFNSGRCLRGVGRDTLSMAIHEVYQELIQDVLKQGYLWKRGHLRRNWAERWFQLQPSCLCYFGSEECKEKRGIIPLDAHCCVEVLPDRDGKRCMFCVKTATRTYEMSASDTRQRQEWTAAIQMAIRLQAEGKTSLHKDLKQKRREQREQR | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction