missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Deerfit Lux Gloves

Product Code. p-7115896
Click to view available options
Size:
8
9
10
11
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12610923

Brand: Honeywell Safety Products™ 200161510

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect your hands from the cold. Honeywell Safety Products™ are best for light and medium handling in cold conditions.

  • Leather glove-fine black suede deerskin.
  • Grip improved by buffed suede grain.
  • Fully lined with Thinsulate™ for cold protection.
  • Breathable, flexible, ultra lightweight, does not absorb humidity.
  • Elasticized wrist for support and reduced thermal exchange.
  • Black color makes the glove more soil proof.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE 1, EC Certificate Number: Autocertification, EC Attestation: EC Attestation
Classification Dexterity: 4
Cut Style Gunn
Color Black
Exterior Finish Buffed Suede
Material Leather, Thinsulate
Product Type Gloves
Size 10
For Use With (Application) Army-Defense, Energy, Electricity, Homeland Defense, Logistics, Service, Telecom, Transportation and Utility industries
Includes Elastic strip
Liner Material Thinsulate™
Lining Lined
Resistant To Cold
Length (Metric) 25 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.