missing translation for 'onlineSavingsMsg'
Learn More

Deepoxy-deoxynivalenol 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325355
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325355 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325355 Supplier Dr. Ehrenstorfer Supplier No. DREV12099000AL50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deepoxy-deoxynivalenol
CAS 88054-24-4
Analyte or Component Names Deepoxy-deoxynivalenol
Shipping Condition Room Temperature
Molecular Formula C15 H20 O5
Concentration 50 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym (3α,7α)-3,7,15-Trihydroxytrichotheca-9,12-dien-8-one, 2,5-Methano-1-benzoxepin, trichotheca-9,12-dien-8-one deriv., DOM 1, Deepoxydeoxynivalenol
InChI Formula InChI=1S/C15H20O5/c1-7-4-10-15(6-16,13(19)11(7)18)14(3)5-9(17)12(20-10)8(14)2/h4,9-10,12-13,16-17,19H,2,5-6H2,1,3H3/t9-,10-,12+,13-,14+,15?/m1/s1
Solution Type Single Solution
SMILES CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)(C3=C)C2(CO)[C@H](O)C1=O
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 280.316
Quantity 5 mL
Formula Weight 280.131 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.