Learn More
Medline Scientific™ Dedicated Platform for BS Series Shaking and Heating Baths
Product Code. p-7138529
Click to view available options
For Use With (Equipment):
BS-06 Bath
BS-11 Bath
BS-21 Bath
BS-31 Bath
Holds:
12 250 mL clamps for BS-31
12 500 mL clamps for BS-31
2 250 mL clamps for BS-06
2 500 mL clamps for BS-06
4 250 mL clamps for BS-11
4 500 mL clamps for BS-11
6 500 mL clamps for BS-21
8 250 mL clamps for BS-21
Unit Size:
Each
Product Code. 15502997
Brand: Medline Scientific™ AAA44542
Only null left
Add to basket
Add to basket
Provides maximum capacity for each platform size when using glassware of the same size.
  • With preinstalled 250 or 500mL clamps
  • For use with BS series Shaking and Heating Baths
Specifications
Description Dedicated platform
For Use With (Equipment) BS-11 Bath
Holds 4 500 mL clamps for BS-11