missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEDD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583152
This item is not returnable.
View return policy
Description
Immunogen sequence: RGRATLGSQR KRRKSVTPDP KEKQTCDIRL RVRAEYCQHE TALQGNVFSN KQDPLERQFE RFNQANTILK SRDLGSIICD IKFSELTYLD AFWRDYINGS LLEALKGVFI TDSLKQAVGH EAIKLLVNV Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
DEDD encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene.
Specifications
| DEDD | |
| Polyclonal | |
| Unconjugated | |
| DEDD | |
| CASP8IP1; death effector domain containing; death effector domain-containing; death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDD; DEDD1; Dedpro1; DEFT; FLDED1; FLDED-1; KE05; tumor necrosis factor (ligand) superfamily, member 19 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9191 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75618 | |
| DEDD | |
| Recombinant protein corresponding to Human DEDD. Recombinant protein control fragment (Product #RP-104613). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction