missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEDD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31686
This item is not returnable.
View return policy
Description
DEDD Polyclonal specifically detects DEDD in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DEDD | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| DEDD | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human DEDD. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (100%) and Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.42mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| CASP8IP1, death effector domain containing, death effector domain-containing, death effector domain-containing protein, Death effector domain-containing testicular molecule, DEDD1, DEDPRO1, DEFTKE05, FLDED1, FLDED-1 | |
| Rabbit | |
| 37 kDa | |
| 0.1 mL | |
| Apoptosis | |
| 9191 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction